KEY184992030 Key Organics / BIONET

2-bromo-5-methoxy-4-methylaniline

  • MDL number MFCD11841053
  • Cas Number : 532440-88-3
  • Molecular Weight : 216.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 75 - 77
Iupac Name 2-bromo-5-methoxy-4-methylaniline
Inchi Code 1S/C8H10BrNO/c1-5-3-6(9)7(10)4-8(5)11-2/h3-4H,10H2,1-2H3
Inchi Key WGVJMLYQPNCPSD-UHFFFAOYSA-N

Description

Melting Point 75 - 77
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106719

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/NC-0740_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart