KEY187620287 Key Organics / BIONET

tert-butyl 2-(hydroxymethyl)-4H,5H,6H,7H,8H-pyrazolo[1,5-a][1,4]diazepine-5-carboxylate

  • MDL number MFCD12025827
  • Cas Number : 1135282-77-7
  • Molecular Weight : 267.329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55 - 57
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3508

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0603_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart