KEY187620291 Key Organics / BIONET

4-(morpholin-4-yl)benzohydrazide

  • MDL number MFCD12025839
  • Cas Number : 1135282-82-4
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 198
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94528

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0295_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart