KEY187620294 Key Organics / BIONET

3-tert-butyl-5-(4-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD11840954
  • Cas Number : 1135282-84-6
  • Molecular Weight : 247.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 144
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94533

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0700_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart