KEY187620300 Key Organics / BIONET

(2E)-3-(5-nitro-1-benzofuran-2-yl)prop-2-enoic acid

  • MDL number MFCD11840959
  • Cas Number : 1135282-87-9
  • Molecular Weight : 233.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292 - 294
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94546

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart