KEY187620303 Key Organics / BIONET

6-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD07374363
  • Cas Number : 367500-94-5
  • Molecular Weight : 180.138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259 - 260
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94550

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart