KEY187620306 Key Organics / BIONET

methyl 4-propanoyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD11840964
  • Cas Number : 1135282-93-7
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-propionyl-1H-pyrrole-2-carboxylate
Inchi Code 1S/C9H11NO3/c1-3-8(11)6-4-7(10-5-6)9(12)13-2/h4-5,10H,3H2,1-2H3
Inchi Key GMDIDIYLWBUNEW-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94553

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AD-0725_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart