KEY187620307 Key Organics / BIONET

1-bromo-2-fluoro-4-methoxy-5-methylbenzene

  • MDL number MFCD11840965
  • Cas Number : 901231-33-2
  • Molecular Weight : 219.053

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 @ 0.2 mm Hg
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94555

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0727_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart