KEY187620308 Key Organics / BIONET

[6-(trifluoromethyl)-1-benzothiophen-2-yl]methanol

  • MDL number MFCD11840966
  • Cas Number : 863118-46-1
  • Molecular Weight : 232.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94558

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0734_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart