KEY187620310 Key Organics / BIONET

4-(3-tert-butyl-1,2,4-oxadiazol-5-yl)benzonitrile

  • MDL number MFCD11840967
  • Cas Number : 1135282-95-9
  • Molecular Weight : 227.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145 - 147
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94564

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0743_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart