KEY187620327 Key Organics / BIONET

5-chloro-2-methyl-6-(methylamino)pyrimidin-4-ol

  • MDL number MFCD12025847
  • Cas Number : 1135283-07-6
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254 - 256
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94598

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0791_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart