KEY187620329 Key Organics / BIONET

1-(4-nitrobenzoyl)-1,2-diazepane

  • MDL number MFCD11840984
  • Cas Number : 1135283-09-8
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82 - 86
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94601

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0795_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart