KEY187620331 Key Organics / BIONET

5-chloro-6-(dimethylamino)-2-methylpyrimidin-4-ol

  • MDL number MFCD11840986
  • Cas Number : 1135283-12-3
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231 - 233
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94603

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0797_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart