KEY187620332 Key Organics / BIONET

ethyl 1-({[(tert-butoxy)carbonyl](2-chloroethyl)amino}sulfonyl)piperidine-4-carboxylate

  • MDL number MFCD12025848
  • Cas Number : 1135283-14-5
  • Molecular Weight : 398.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82 - 83
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94605

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-2002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart