KEY187620337 Key Organics / BIONET

3-chloro-2-ethynyl-5-(trifluoromethyl)pyridine

  • MDL number MFCD11840989
  • Cas Number : 1135283-18-9
  • Molecular Weight : 205.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 24 @ 0.1 mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94612

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-5008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart