KEY187620342 Key Organics / BIONET

3-[4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile

  • MDL number MFCD07787044
  • Cas Number : 887591-40-4
  • Molecular Weight : 230.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95030

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BD-0200_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart