KEY187620368 Key Organics / BIONET

1-benzyl-2-methyl-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD04539177
  • Cas Number : 14624-97-6
  • Molecular Weight : 237.306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112 - 114
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94604

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0799_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart