KEY187620373 Key Organics / BIONET

5-nitro-2-(piperidin-1-yl)aniline

  • MDL number MFCD00852821
  • Cas Number : 5367-58-8
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-nitro-2-(1-piperidinyl)aniline
Inchi Code 1S/C11H15N3O2/c12-10-8-9(14(15)16)4-5-11(10)13-6-2-1-3-7-13/h4-5,8H,1-3,6-7,12H2
Inchi Key DMIMWGHYIPFAIF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94537

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AD-0704_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart