KEY187620374 Key Organics / BIONET

N-(2-fluoro-5-nitrophenyl)methanesulfonamide

  • MDL number MFCD01213923
  • Cas Number : 123343-99-7
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 165
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94567

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0746_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart