KEY187620376 Key Organics / BIONET

7-fluoro-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD07774200
  • Cas Number : 103361-99-5
  • Molecular Weight : 167.139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203 - 205
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94523

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AD-0189_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart