KEY187620377 Key Organics / BIONET

2-(morpholin-4-yl)-5-nitroaniline

  • MDL number MFCD02575506
  • Cas Number : 4031-79-2
  • Molecular Weight : 223.232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152 - 154
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94539

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart