KEY187620379 Key Organics / BIONET

2-(2-fluoro-5-nitrophenyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD01119713
  • Cas Number : 166658-69-1
  • Molecular Weight : 286.218

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94572

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0753_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart