KEY187620383 Key Organics / BIONET

2-methyl-6-(methylamino)pyrimidin-4-ol

  • MDL number MFCD11840972
  • Cas Number : 857414-73-4
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 288 - 291
Iupac Name 2-methyl-6-(methylamino)-4-pyrimidinol
Inchi Code 1S/C6H9N3O/c1-4-8-5(7-2)3-6(10)9-4/h3H,1-2H3,(H2,7,8,9,10)
Inchi Key SYONBPZOUULYEZ-UHFFFAOYSA-N

Description

Melting Point 288 - 291
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94574

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AD-0756_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart