KEY187620384 Key Organics / BIONET

(2E)-3-[2-(morpholin-4-yl)-5-nitrophenyl]prop-2-enoic acid

  • MDL number MFCD06205585
  • Cas Number : 300665-18-3
  • Molecular Weight : 278.264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94575

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0758_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart