KEY187620385 Key Organics / BIONET

[4'-(1,3-dioxolan-2-yl)-[1,1'-biphenyl]-4-yl]methanol

  • MDL number MFCD06802587
  • Cas Number : 889954-12-5
  • Molecular Weight : 256.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 - 125
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94578

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0761_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart