KEY187620389 Key Organics / BIONET

2-(4-chlorophenyl)-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11840957
  • Cas Number : 187998-87-4
  • Molecular Weight : 307.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 182
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94543

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0712_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart