KEY187620396 Key Organics / BIONET

5-bromo-2-ethoxypyridine-3-carboxylic acid

  • MDL number MFCD09836166
  • Cas Number : 393184-78-6
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 123
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76407

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0072_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart