KEY187620398 Key Organics / BIONET

1-benzyl-5-nitro-1H-1,3-benzodiazole

  • MDL number MFCD00837324
  • Cas Number : 15207-93-9
  • Molecular Weight : 253.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94587

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0771_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart