KEY187620399 Key Organics / BIONET

5,6-dichloro-2-methylpyrimidin-4-ol

  • MDL number MFCD12025845
  • Cas Number : 32265-50-2
  • Molecular Weight : 179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 249 - 251
Iupac Name 5,6-dichloro-2-methyl-4-pyrimidinol
Inchi Code 1S/C5H4Cl2N2O/c1-2-8-4(7)3(6)5(10)9-2/h1H3,(H,8,9,10)
Inchi Key KUQPBCZFPWRAHB-UHFFFAOYSA-N

Description

Melting Point 249 - 251
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94588

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AD-0773_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart