KEY187620422 Key Organics / BIONET

4-(4-methylpiperazin-1-yl)benzoic acid hydrochloride

  • MDL number MFCD06641474
  • Cas Number : 289044-60-6
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245 (dec)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94530

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0334_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart