KEY187620425 Key Organics / BIONET

4-hydrazinyl-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD01223746
  • Cas Number : 104408-29-9
  • Molecular Weight : 156.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143 - 145
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106726

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NC-0750_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart