KEY187620428 Key Organics / BIONET

6-fluoroimidazo[1,2-a]pyridine

  • MDL number MFCD09994632
  • Cas Number : 139022-27-8
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 70 - 71
Iupac Name 6-fluoroimidazo[1,2-a]pyridine
Inchi Code 1S/C7H5FN2/c8-6-1-2-7-9-3-4-10(7)5-6/h1-5H
Inchi Key ORLNGHHPODFQRV-UHFFFAOYSA-N

Description

Melting Point 70 - 71
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106731

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/NC-0755_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart