KEY187620431 Key Organics / BIONET

tert-butyl N-[2-(morpholin-4-yl)-2-oxoethyl]carbamate

  • MDL number MFCD07366739
  • Cas Number : 114703-81-0
  • Molecular Weight : 244.291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88247

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Y-0617_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart