KEY189464689 Key Organics / BIONET

1-methyl-3-(trifluoromethyl)-5-[2-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD12025830
  • Cas Number : 1176437-32-3
  • Molecular Weight : 338.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84518

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Z-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart