KEY189464693 Key Organics / BIONET

3,5-diethyl 4-chloropyridine-3,5-dicarboxylate

  • MDL number MFCD12025855
  • Cas Number : 244638-43-5
  • Molecular Weight : 257.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95028

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BD-0110_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart