KEY189464712 Key Organics / BIONET

ethyl 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

  • MDL number MFCD12025935
  • Cas Number : 128073-39-2
  • Molecular Weight : 257.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97604

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/H-20242_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart