KEY189464713 Key Organics / BIONET

2-nitro-5-(piperidin-1-yl)benzoic acid

  • MDL number MFCD09787496
  • Cas Number : 118159-39-0
  • Molecular Weight : 250.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 163
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95053

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BD-0716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart