KEY189464716 Key Organics / BIONET

6-methylpyrimidine-2,4-diamine

  • MDL number MFCD00127861
  • Cas Number : 1791-73-7
  • Molecular Weight : 124.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196 - 198 C
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95062

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BD-0732_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart