KEY189464722 Key Organics / BIONET

ethyl 2-amino-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate

  • MDL number MFCD09909738
  • Cas Number : 135026-17-4
  • Molecular Weight : 224.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188 - 190 C
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95667

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CD-0201_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart