KEY189464726 Key Organics / BIONET

3,5-diethyl 4-oxo-1,4-dihydropyridine-3,5-dicarboxylate

  • MDL number MFCD12025854
  • Cas Number : 74632-03-4
  • Molecular Weight : 239.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 4-oxo-1,4-dihydro-3,5-pyridinedicarboxylate
Inchi Code 1S/C11H13NO5/c1-3-16-10(14)7-5-12-6-8(9(7)13)11(15)17-4-2/h5-6H,3-4H2,1-2H3,(H,12,13)
Inchi Key GGMYLXLRJKRWQM-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95024

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BD-0078_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart