KEY189464728 Key Organics / BIONET

2,3,5-trichloropyridine-4-carboxylic acid

  • MDL number MFCD11044390
  • Cas Number : 406676-18-4
  • Molecular Weight : 226.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 193 - 194 C
Iupac Name 2,3,5-trichloroisonicotinic acid
Inchi Code 1S/C6H2Cl3NO2/c7-2-1-10-5(9)4(8)3(2)6(11)12/h1H,(H,11,12)
Inchi Key TYMTYHMQJHSRIF-UHFFFAOYSA-N

Description

Melting Point 193 - 194 C
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95678

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CD-0714_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart