KEY189464733 Key Organics / BIONET

(E)-N-[(1-methyl-1H-pyrrol-2-yl)methylidene]hydroxylamine

  • MDL number MFCD09054061
  • Cas Number : 37110-15-9
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 146 - 148
Iupac Name 1-methyl-1H-pyrrole-2-carbaldehyde oxime
Inchi Code 1S/C6H8N2O/c1-8-4-2-3-6(8)5-7-9/h2-4H,5H2,1H3
Inchi Key ZBSBDBRMWHWWBN-UHFFFAOYSA-N

Description

Melting Point 146 - 148
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87070

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7P-087_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart