KEY189464734 Key Organics / BIONET

2-methoxy-4,6-dimethylpyridine-3-carbonitrile

  • MDL number MFCD00124716
  • Cas Number : 65515-39-1
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methoxy-4,6-dimethylnicotinonitrile
Inchi Code 1S/C9H10N2O/c1-6-4-7(2)11-9(12-3)8(6)5-10/h4H,1-3H3
Inchi Key WEJLKGQPSKTCBR-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95684

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CD-0725_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart