KEY189464738 Key Organics / BIONET

ethyl 4-(4-aminophenyl)piperazine-1-carboxylate

  • MDL number MFCD11848963
  • Cas Number : 16154-70-4
  • Molecular Weight : 249.314

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66352

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0601_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart