KEY195583051 Key Organics / BIONET

{[(3Z)-2-oxo-2,3-dihydro-1H-indol-3-ylidene]amino}thiourea

  • MDL number MFCD00043581
  • Cas Number : 487-16-1
  • Molecular Weight : 220.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252 - 254
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6842

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-1254_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart