KEY195583053 Key Organics / BIONET

ethyl (2E)-2-chloro-2-[2-(4-methoxy-2-nitrophenyl)hydrazin-1-ylidene]acetate

  • MDL number MFCD06200941
  • Cas Number : 119750-09-3
  • Molecular Weight : 301.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66556

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0960_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart