KEY195583059 Key Organics / BIONET

ethyl (2E)-2-[2-(2,5-dichlorophenyl)hydrazin-1-ylidene]-2-acetamidoacetate

  • MDL number MFCD08056615
  • Cas Number : 886361-49-5
  • Molecular Weight : 318.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 192
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67286

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-338S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart