KEY195583060 Key Organics / BIONET

(3Z)-3-[2-(2,4,6-trichlorophenyl)hydrazin-1-ylidene]-2,3-dihydro-1H-indole-2-thione

  • MDL number MFCD06200943
  • Cas Number : 862665-28-9
  • Molecular Weight : 356.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254 - 255
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69215

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-1255_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart