KEY195583063 Key Organics / BIONET

(E)-{[2-(benzyloxy)phenyl]methylidene}(methyl)oxidoazanium

  • MDL number MFCD03001260
  • Cas Number : 338422-69-8
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60 - 62
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74815

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart