KEY195583065 Key Organics / BIONET

(E)-N'-{6-benzyl-3-cyano-4H,5H,6H,7H-thieno[2,3-c]pyridin-2-yl}-N,N-dimethylmethanimidamide

  • MDL number MFCD11553021
  • Cas Number : 1092352-17-4
  • Molecular Weight : 324.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77496

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Z-0227_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart